missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HOXB1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6318
This item is not returnable.
View return policy
Description
HOXB1 Polyclonal specifically detects HOXB1 in Human samples. It is validated for Immunocytochemistry.
Specifications
| HOXB1 | |
| Polyclonal | |
| Unconjugated | |
| P14653 | |
| HOXB1 | |
| E. coli-derived recombinant human HOXB1 Met1-Thr192 Accession # P14653 | |
| 100 μg | |
| Primary | |
| Detects human HOXB1 in direct ELISAs. In direct ELISAs, less than 2% cross-reactivity with recombinant human (rh) HOXA1, rhHOXB4, and rhHOXA9 is observed. | |
| Human | |
| IgG |
| Immunocytochemistry | |
| LYOPH | |
| Immunocytochemistry 5-15 ug/mL | |
| homeo box 2I, homeo box B1, homeobox B1, Homeobox protein Hox-2I, homeobox protein Hox-B1, HOX2, Hox-2.9, HOX2I, HOX2IMGC116844, MGC116843, MGC116845 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 3211 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction