missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Human HMGB1/HMG-1 ELISA Kit (Chemiluminescence)
Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
The Human HMGB1/HMG-1 ELISA Kit (Chemiluminescence) measures human HMGB1/HMG-1 in serum, plasma and other biological fluids.
Specifications
Specifications
| Assay Range | 12.50-800 pg/mL |
| Assay Sensitivity | 7.50 pg/mL |
| Cross Reactivity | This kit recognizes Human HMGB-1 in samples. No significant cross-reactivity or interference between Human HMGB-1 and analogues was observed. |
| Product Type | ELISA Kit (Chemiluminescence) |
| Sample Type | Biological Fluids, Plasma, Serum |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3146 |
| Gene Symbol | HMGB1 |
| Host Species | Human |
| Quantity | 1 Kit |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction