missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human HMGA1 (aa 61-94) Control Fragment Recombinant Protein

Product Code. 30206220
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30206220 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30206220 Supplier Invitrogen™ Supplier No. RP109158

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

HMGA1 encodes a non-histone protein involved in many cellular processes, including regulation of inducible gene transcription, integration of retroviruses into chromosomes, and the metastatic progression of cancer cells. The encoded protein preferentially binds to the minor groove of A+T-rich regions in double-stranded DNA. It has little secondary structure in solution but assumes distinct conformations when bound to substrates such as DNA or other proteins. The encoded protein is frequently acetylated and is found in the nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P17096
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 3159
Name Human HMGA1 (aa 61-94) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AL023995; HGNC:5010; high mobility group AT-hook 1; high mobility group AT-hook protein 1; high mobility group HMGA1A; high mobility group HMGA1B; high mobility group protein; high mobility group protein 1 A; high mobility group protein A1; high mobility group protein A1A; high mobility group protein A1B; high mobility group protein HMG-I/HMG-Y; high mobility group protein I; high mobility group protein R; high mobiliy group protein A1B; high-mobility group (nonhistone chromosomal) protein isoforms I and Y; HMGA1; Hmga1a; Hmga1b; Hmgi; HMG-I(Y); HMGIY; HMG-R; Hmgy; MGC12816; MGC4242; MGC4854; nonhistone chromosomal high-mobility group protein HMG-I/HMG-Y; non-histone protein; non-histone protein; HMGA1A; non-histone protein; HMGA1B; OTTHUMP00000016222; OTTHUMP00000016223; OTTHUMP00000016224; OTTHUMP00000039618
Common Name HMGA1
Gene Symbol HMGA1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PKGSKNKGAAKTRKTTTTPGRKPRGRPKKLEKEE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.