missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human HLA-DOA (aa 25-74) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (86%), Rat (86%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Reacts with HLA-DO, a heterodimer formed by DNalpha and DObeta subunits in B cells. DNalpha and DObeta are the products of the non-classical class II genes, HLA-DNA and HLA-DOB, respectively. DO forms tight complexes with DM in the endoplasmic reticulum and is thereby sorted to lysosomal vesicles during antigen processing and presentation. It is tightly associated with DM and it is selectively expressed on antigen-presenting cells, such as B cells, dendritic cells and thymic epithelial cells. DO can enhance the efficiency of peptide loading and has been found to stabilize DM at low pH, preserving its chaperon activity. DO-DM complexes are more efficient than DM in protecting empty DR molecules. Reports describe DO as a co-chaperone of DM. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
Specifications
Specifications
| Accession Number | P06340 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 3111 |
| Name | Human HLA-DOA (aa 25-74) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | HLA class II histocompatibility antigen, DO alpha chain; HLA-D0-alpha; HLA-DNA; HLA-DOA; HLADZ; HLA-DZA; lymphocyte antigen; major histocompatibility complex, class II, DN alpha; major histocompatibility complex, class II, DO alpha; MHC class II antigen DOA; MHC DN-alpha; MHC DZ alpha |
| Common Name | HLA-DOA |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction