missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Human HLA Class I Antibody, Novus Biologicals™

Mouse Monoclonal Antibody
£177.00 - £469.00
Specifications
| Antigen | HLA Class I |
|---|---|
| Clone | 1069511 |
| Dilution | Western Blot 2 μg/mL, Flow Cytometry 2.5 μg/106 cells, Immunohistochemistry 3-25 μg/mL |
| Applications | Flow Cytometry, Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30018977
|
Novus Biologicals™
MAB11502-SP |
25 μg |
£177.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30018994
![]() |
Novus Biologicals™
MAB11502-100 |
100 μg |
£469.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HLA Class I Monoclonal antibody specifically detects HLA Class I in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western BlotSpecifications
| HLA Class I | |
| Western Blot 2 μg/mL, Flow Cytometry 2.5 μg/106 cells, Immunohistochemistry 3-25 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. See Certificate of Analysis for details. *Small pack size (-SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| 3105 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze/thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
| 1069511 | |
| Flow Cytometry, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, HLA-A, HLA-ABC, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80 | |
| Membranes from human tonsillar lymphocytes, | |
| Primary | |
| Reconstitute lyophilized material at 0.2mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title