missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Histone H2AX Antibody, Novus Biologicals™
Mouse Monoclonal Antibody
£189.00 - £429.00
Specifications
| Antigen | Histone H2AX |
|---|---|
| Clone | 1077751 |
| Dilution | Immunohistochemistry 3-25 μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30114380
|
R&D Systems
MAB11529-100 |
100 μg |
£429.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30116610
|
R&D Systems
MAB11529-SP |
25 μg |
£189.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Histone H2AX Monoclonal antibody specifically detects Histone H2AX in Human samples. It is validated for ImmunohistochemistrySpecifications
| Histone H2AX | |
| Immunohistochemistry 3-25 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. See Certificate of Analysis for details. *Small pack size (-SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| H2A.X, H2A/X, H2AFX | |
| E. coli -derived recombinant human H2AX, Gly3-Ser131, Accession # P16104 | |
| Primary | |
| Reconstitute lyophilized material at 0.2mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. |
| 1077751 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P16104 | |
| 3014 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title