missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Histamine N-Methyltransferase/HNMT Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7637-SP
This item is not returnable.
View return policy
Description
Histamine N-Methyltransferase/HNMT Polyclonal specifically detects Histamine N-Methyltransferase/HNMT in Human samples. It is validated for Western Blot.
Specifications
| Histamine N-Methyltransferase/HNMT | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.1.1.8, histamine N-methyltransferase, Histamine NMethyltransferase, HMT, HNMT, HNMT-S1, HNMT-S2 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 3176 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| P50135 | |
| HNMT | |
| E. coli-derived recombinant human Histamine N-Methyltransferase/HNMT Ser7-Ala292 Accession # P50135 | |
| 25 μg | |
| Primary | |
| Detects human Histamine N-Methyltransferase/HNMT in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion