missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human HIST2H2BE (aa 35-60) Control Fragment Recombinant Protein

Product Code. 30206240
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30206240

Brand: Invitrogen™ RP104350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60199 (PA5-60199. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q16778
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 8349
Name Human HIST2H2BE (aa 35-60) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AV127319; GL105; H2B; H2b 613; H2B histone family, member Q; H2B0.1; H2B/q; H2b-613; H2BFQ; H2BGL105; H2BQ; Hist2h2be; HIST2H2BF; histone 2, H2be; histone cluster 2, H2be; histone cluster 2, H2bf; histone H2B type 2-E; Histone H2B type 2-F; Histone H2B.q; histone H2B-GL105; Histone H2Bq; RGD1562346; T25626
Common Name HIST2H2BE
Gene Symbol H2BC21
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KESYSIYVYKVLKQVHPDTGISSKAM
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.