missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human High Sensitivity Cytokine B Premixed Magnetic Luminex™ Performance Assay

Product Code. 18401420
Change view
Click to view available options
Quantity:
10 Plex
11 Plex
12 Plex
13 Plex
14 Plex
15 Plex
16 Plex
17 Plex
2 Plex
3 Plex
4 Plex
5 Plex
6 Plex
7 Plex
8 Plex
9 Plex
Unit Size:
1 set
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18401420 4 Plex 1 set
18461310 11 Plex 1 set
18442400 6 Plex 1 set
18411440 8 Plex 1 set
18462090 7 Plex 1 set
18401520 5 Plex 1 set
18412330 3 Plex 1 set
18492320 2 Plex 1 set
18432330 9 Plex 1 set
18402300 10 Plex 1 set
18461150 14 Plex 1 set
18451810 15 Plex 1 set
18412210 16 Plex 1 set
18471880 17 Plex 1 set
18442430 12 Plex 1 set
18401740 13 Plex 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
This item is not returnable. View return policy
Product Code. 18401420 Supplier R&D Systems™ Supplier No. FCSTM1404

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Magnetic bead-based multiplex assay for the Luminex™ platform

TRUSTED_SUSTAINABILITY

Specifications

Quantity 4 Plex
Species Human
For Use With (Application) ELISA
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.