missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human High Sensitivity Cytokine A Premixed Magnetic Luminex™ Performance Assay

Product Code. 18421440
Change view
Click to view available options
Quantity:
10 Plex
11 Plex
12 Plex
2 Plex
3 Plex
4 Plex
5 Plex
6 Plex
7 Plex
8 Plex
9 Plex
Unit Size:
1 set
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421440 11 Plex 1 set
18401930 6 Plex 1 set
18461920 2 Plex 1 set
18432520 3 Plex 1 set
18492290 4 Plex 1 set
18412400 5 Plex 1 set
18462520 8 Plex 1 set
18411970 7 Plex 1 set
18481630 9 Plex 1 set
18402130 10 Plex 1 set
18462210 12 Plex 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 18421440 Supplier R&D Systems™ Supplier No. FCSTM0911

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Magnetic bead-based multiplex assay for the Luminex™ platform

TRUSTED_SUSTAINABILITY

Specifications

Quantity 11 Plex
Species Human
For Use With (Application) ELISA
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.