missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HGK Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF6027-SP
This item is not returnable.
View return policy
Description
HGK/MAP4K4 Polyclonal specifically detects HGK/MAP4K4 in Human samples. It is validated for Western Blot.
Specifications
| HGK/MAP4K4 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.7.11, EC 2.7.11.1, FLH21957, FLJ10410, hepatocyte progenitor kinase-like/germinal center kinase-like kinase, HGKFLJ20373, HPK/GCK-like kinase HGK, KIAA0687, MAP4K4, MAPK/ERK kinase kinase kinase 4, MEK kinase kinase 4, MEKKK 4, MEKKK4, mitogen-activated protein kinase kinase kinase kinase 4, Nck-interacting kinase, NIKFLJ90111 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 9448 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| O95819-4 | |
| MAP4K4 | |
| E. coli-derived recombinant human HGK Ser686-Val820 Accession # O95819-4 | |
| 25 μg | |
| Primary | |
| Detects human HGK in Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction