missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HGFR/c-MET Antibody, Novus Biologicals™

Rabbit Monoclonal Antibody
£169.00 - £492.00
Specifications
| Antigen | HGFR/c-MET |
|---|---|
| Clone | 2504B |
| Dilution | Immunocytochemistry 3-25 ug/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18795775
![]() |
R&D Systems
MAB11320-100 |
100 μg |
£492.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18795935
![]() |
R&D Systems
MAB11320-SP |
25 μg |
£169.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HGFR/c-MET Monoclonal antibody specifically detects HGFR/c-MET in Human samples. It is validated for ImmunocytochemistrySpecifications
| HGFR/c-MET | |
| Immunocytochemistry 3-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| AUTS9, c-Met, cMET, EC 2.7.10, EC 2.7.10.1, hepatocyte growth factor receptor, HGF R, HGF receptor, HGF/SF receptor, HGFR, MET, Met (c-Met), met proto-oncogene (hepatocyte growth factor receptor), met proto-oncogene tyrosine kinase, oncogene MET, Proto-oncogene c-Met, RCCP2, Scatter factor receptor, SF receptor, Tyrosine-protein kinase Met | |
| Synthetic peptide HGFR_Y1234, Accession # P08581 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 2504B | |
| Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P08581 | |
| 4233 | |
| IgG | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title