missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HGF Activator Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1514-SP
This item is not returnable.
View return policy
Description
HGF Activator Polyclonal specifically detects HGF Activator in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
Specifications
| HGF Activator | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| HGF activator, HGFA, Hgfac | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3083 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q04756.1 | |
| HGFAC | |
| Mouse myeloma cell line NS0-derived recombinant human HGF Activator Gln36-Ser655 Accession # Q04756.1 | |
| 25 μg | |
| Primary | |
| Detects human HGF Activator in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant mouse HGF Activator is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction