missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Hexosaminidase A/HEXA Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB62371
This item is not returnable.
View return policy
Description
Hexosaminidase A/HEXA Monoclonal specifically detects Hexosaminidase A/HEXA in Human samples. It is validated for Neutralization.
Specifications
| Hexosaminidase A/HEXA | |
| Monoclonal | |
| LYOPH | |
| Neutralization 3.0 ug/mL | |
| P06865 | |
| HEXA | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Hexosaminidase A/HEXA Met1-Thr529 Accession # P06865 | |
| 100 μg | |
| Primary | |
| Detects human Hexosaminidase A/HEXA in direct ELISAs. | |
| Human | |
| Purified |
| Neutralization | |
| 714712 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| beta-hexosaminidase subunit alpha, Beta-N-acetylhexosaminidase subunit alpha, EC 3.2.1, EC 3.2.1.52, HEXA, hexosaminidase A (alpha polypeptide), Hexosaminidase subunit A, MGC99608, N-acetyl-beta-glucosaminidase subunit alpha, TSD | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3073 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction