missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Hexim 1 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF8106
This item is not returnable.
View return policy
Description
Hexim 1 Polyclonal specifically detects Hexim 1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Hexim 1 | |
| Polyclonal | |
| Unconjugated | |
| O94992 | |
| HEXIM1 | |
| E. coli-derived recombinant human Hexim 1 His213-Asp359 Accession # O94992 | |
| 100 μg | |
| Primary | |
| Detects human Hexim 1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human Hexim 2 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Cardiac lineage protein 1, CLP1, CLP-1, CLP1hexamethylene-bis-acetamide-inducible transcript 1, EDG1HMBA-inducible, Estrogen down-regulated gene 1 protein, hexamethylene bis-acetamide inducible 1, Hexamethylene bis-acetamide-inducible protein 1, hexamthylene bis-acetamide inducible 1, HEXIM1, HIS1, HIS1FLJ13562, HMBA-inducible, MAQ1, MAQ1hexamethylene bisacetamide-inducible protein, menage a quatre 1, Menage a quatre protein 1, protein HEXIM1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 10614 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction