missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Hexim 1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF8106-SP
This item is not returnable.
View return policy
Description
Hexim 1 Polyclonal specifically detects Hexim 1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Hexim 1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cardiac lineage protein 1, CLP1, CLP-1, CLP1hexamethylene-bis-acetamide-inducible transcript 1, EDG1HMBA-inducible, Estrogen down-regulated gene 1 protein, hexamethylene bis-acetamide inducible 1, Hexamethylene bis-acetamide-inducible protein 1, hexamthylene bis-acetamide inducible 1, HEXIM1, HIS1, HIS1FLJ13562, HMBA-inducible, MAQ1, MAQ1hexamethylene bisacetamide-inducible protein, menage a quatre 1, Menage a quatre protein 1, protein HEXIM1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 10614 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| O94992 | |
| HEXIM1 | |
| E. coli-derived recombinant human Hexim 1 His213-Asp359 Accession # O94992 | |
| 25 μg | |
| Primary | |
| Detects human Hexim 1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human Hexim 2 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction