missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Heparan Sulfate 6-O-Sulfotransferase 1/HS6ST1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5057-SP
This item is not returnable.
View return policy
Description
Heparan Sulfate 6-O-Sulfotransferase 1/HS6ST1 Monoclonal specifically detects Heparan Sulfate 6-O-Sulfotransferase 1/HS6ST1 in Human samples. It is validated for Immunohistochemistry.
Specifications
| Heparan Sulfate 6-O-Sulfotransferase 1/HS6ST1 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| O60243 | |
| HS6ST1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Heparan Sulfate 6-O-Sulfotransferase 1/HS6ST1 Pro38-Trp411 Accession # O60243 | |
| 25 μg | |
| Primary | |
| Detects human Heparan Sulfate 6-O-Sulfotransferase 1/HS6ST1 in direct ELISAs. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 584308 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp547H098, EC 2.8.2, EC 2.8.2.-, FLJ25392, heparan sulfate 6-O-sulfotransferase 1, heparan-sulfate 6-sulfotransferase, HS6ST-1, HS6STheparan-sulfate 6-O-sulfotransferase 1, MGC116899, MGC116901 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9394 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction