missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Heparan Sulfate 2-O-Sulfotransferase 1/HS2ST1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB6335
This item is not returnable.
View return policy
Description
Heparan Sulfate 2-O-Sulfotransferase 1/HS2ST1 Monoclonal specifically detects Heparan Sulfate 2-O-Sulfotransferase 1/HS2ST1 in Human samples. It is validated for Western Blot.
Specifications
| Heparan Sulfate 2-O-Sulfotransferase 1/HS2ST1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| Q7LGA3 | |
| HS2ST1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Heparan Sulfate 2-O-Sulfotransferase 1/HS2ST1 Met59-Asn356 Accession # Q7LGA3 | |
| 100 μg | |
| Primary | |
| Detects human Heparan Sulfate 2-O-Sulfotransferase 1/HS2ST1 in ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 738306 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 2OST, 2-O-sulfotransferase, EC 2.8.2, EC 2.8.2.-, FLJ11317, heparan sulfate 2-O-sulfotransferase 1, HS2ST, KIAA0448dJ604K5.2, MGC131986 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9653 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction