missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Human Heart Whole Tissue Lysate (Adult Whole Hypertension)
Shop All Bio Techne ProductsDescription
The Human Heart Whole Tissue Lysate (Adult Whole Hypertension) is derived from human.
Specifications
Specifications
| Tissue | Heart |
| Quantity | 1 mg |
| Storage Buffer | HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors |
| Test Specificity | Total Protein - hypertension: Artery |
| Cellular Fraction | Whole |
| Tissue Condition | Hypertension |
| Cross Reactivity | Human |
| Host Species | Human |
| Product Type | Human Heart Whole Tissue Lysate (Adult Hypertension) |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?