missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human HDJ2 (aa 244-379) Control Fragment Recombinant Protein

Product Code. 30195413
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30195413

Brand: Invitrogen™ RP95148

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-81929 (PA5-81929. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1. 6 kb and a protein product with a molecular weight of about 35 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P31689
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 3301
Name Human HDJ2 (aa 244-379) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias DJ-2; DjA1; DnaJ (Hsp40) homolog, subfamily A, member 1; DnaJ heat shock protein family (Hsp40) member A1; dnaJ homolog subfamily A member 1; DnaJ protein homolog 2; Dnaj2; DNAJA1; dnaJ-like protein 1; DnaJ-like protein 2; DnaJ-like subfamily A member 1; h HSJ-2; HDJ2; hDj-2; Heat shock 40 kDa protein 4; heat shock protein J2; heat shock protein, DNAJ-like 2; HSDJ; HSJ2; HSJ-2; HSPF4; human DnaJ protein 2; I79_019991; LOC100346508; LOW QUALITY PROTEIN: dnaJ homolog subfamily A member 1; NEDD7; neural precursor cell expressed, developmentally down-regulated 7; neural precursor cell expressed, developmentally down-regulated gene 7; Rdj1
Common Name HDJ2
Gene Symbol DNAJA1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DQKDHAVFTRRGEDLFMCMDIQLVEALCGFQKPISTLDNRTIVITSHPGQIVKHGDIKCVLNEGMPIYRRPYEKGRLIIEFKVNFPENGFLSPDKLSLLEKLLPERKEVEETDEMDQVELVDFDPNQERRRHYNGE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.