missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human HDAC4 (aa 1071-1084) Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Human HDAC4 (residues 1071-1084) Synthetic Peptide, PEP-141, from Invitrogen.
Histone deacetylase 4 (HDAC4) is responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3, and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. HDAC4 is also involved in muscle maturation via its interaction with the myocyte enhancer factors such as MEF2A, MEF2C and MEF2D. It is also involved in the MTA1-mediated epigenetic regulation of ESR1 expression in breast cancer. Mutations to the HDAC4 gene have been found in Brachydactyly-mental retardation syndrome (BDMR).
Specifications
Specifications
| Gene ID (Entrez) | 9759 |
| Species | Human |
| Purification Method | purified |
| Accession Number | P56524 |
| Common Name | Human HDAC4 (aa 1071-1084) |
| Concentration | 0.5 mg/mL |
| Content And Storage | -20°C. Avoid freeze-thaw cycles. |
| Description | Synthetic Peptide |
| Format | Lyophilized |
| Formulation | peptide with no preservative |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction