missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human HCC-1 (CCL14) (66 aa) Recombinant Protein, PeproTech®

Product Code. 17882633
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
500 μg
missing translation for 'unitSize'
100µg
10µg
1mg
250µg
2µg
500µg
This item is not returnable. View return policy

Product Code. 17882633

missing translation for 'mfr': Gibco™ 30038B2UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human HCC-1 (66 a.a.) is a 7.8 kDa protein consisting of 66 amino acids, including the four highly conserved residues present in CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This gene, CCL14, is one of several CC cytokine genes clustered on 17q11.2. The CC cytokines are secreted proteins characterized by two adjacent cysteines. The cytokine encoded by this gene induces changes in intracellular calcium concentration and enzyme release in monocytes. This gene expresses both monocistronic and bicistronic transcripts. Bicistronic transcripts include the upstream cytokine gene CCL15. In addition, CCL14 undergoes alternative splicing in the bicistronic transcripts; it is unknown if its monocistronic transcript is also subject to alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q16627
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 6358
Molecular Weight (g/mol) 7.8 kDa
Name Human HCC-1 (CCL14) (66 aa)
Quantity 2 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias C-C motif chemokine 14; C-C motif chemokine ligand 14; CC-1; CC-3; CCL14; chemokine (C-C motif) ligand 14; chemokine CC-1/CC-3; chemokine CC-3; CKB1; H HCC-1(1-74); HCC-1; HCC-1(1-74); HCC-1(3-74); HCC-1(4-74); HCC-1(9-74); HCC-1/HCC-3; HCC-3; hemofiltrate CC chemokine 1; MCIF; NCC2; NCC-2; new CC chemokine 2; SCYA14; SCYL2; small inducible cytokine subfamily A (Cys-Cys), member 14; small-inducible cytokine A14; SY14
Common Name CCL14
Gene Symbol CCL14
Biological Activity Determined by its ability to chemoattract human monocytes using a concentration range of 5.0-20.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence GPYHPSECCF TYTTYKIPRQ RIMDYYETNS QCSKPGIVFI TKRGHSVCTN PSDKWVQDYI KDMKEN
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.