missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human HBB Recombinant Protein

Product Code. 11548901
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11548901 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11548901 Supplier Invitrogen™ Supplier No. RP75575

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

RP-75575 contains purified recombinant human Haptoglobin b-Chain protein with an N-terminal hexahistidine tag. RP-75575 may be used as a positive control in many immunological assays.

The alpha (HBA) and beta (HBB) loci determine the structure of the 2 types of polypeptide chains in adult hemoglobin, Hb A. The normal adult hemoglobin tetramer consists of two alpha chains and two beta chains. Mutant beta globin causes sickle cell anemia. Absence of beta chain causes beta-zero-thalassemia. Reduced amounts of detectable beta globin causes beta-plus-thalassemia. The order of the genes in the beta-globin cluster is 5'-epsilon-gamma-G-gamma-A-delta-beta-3'.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P68871
Concentration 0.05 mg/mL
For Use With (Application) Control, Western Blot
Formulation Laemmli with no preservative; pH 6.8
Gene ID (Entrez) 3043
Name Human HBB
pH Range 6.8
Quantity 10 μg
Source E. coli
Immunogen N/A
Regulatory Status RUO
Gene Alias beta globin chain; beta-1-globin; Beta-globin; CD113t-C; Hbb; hemoglobin beta; Hemoglobin beta chain; hemoglobin beta chain complex; hemoglobin beta chain, major-form; Hemoglobin beta-1 chain; hemoglobin subunit beta; Hemoglobin subunit beta-1; hemoglobin, beta; III beta-1 globin; III beta-2 globin; LVV-hemorphin-7; Spinorphin
Common Name HBB
Gene Symbol HBB
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Protein Tag His-tag
Sequence Amino acids Ile162- Asn406 containing an N-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.