missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human HAT (NP_004253.1) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Specifications
Specifications
| Gene | Airway Trypsin-like Protease/HAT/TMPRSS11D |
| Accession Number | NP_004253.1 |
| Gene ID (Entrez) | 9407 |
| Gene Alias | airway trypsin like protease, Airway trypsin-like protease, EC 3.4.21, EC 3.4.21.-, HAT, MGC150587, MGC150588, TMPRSS11D, transmembrane protease serine 11D, transmembrane protease, serine 11D |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction