missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human HAPLN4 (NP_075378) VersaClone cDNA

Product Code. 18715299
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18715299

Brand: R&D Systems™ RDC2598

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sequence verified insert equipped with translated sequences for the insert and entire plasmid DNA.

  • The Human HAPLN4 (NP_075378) VersaClone cDNA clone contains the complete ORF for HAPLN4, along with a Kozak consensus sequence for optimal translation initiation
  • The gene insert is flanked with convenient multiple cloning sites, which can be used to easily cut and transfer the gene cassette into your desired expression vector
  • The HAPLN4 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
TRUSTED_SUSTAINABILITY

Specifications

Gene hHAPLN4
Accession Number NP_075378
Concentration 10 μg at 0.2 μg/μL
Formulation DNA is provided in 10 mM Tris-Cl, pH 8.5
Gene ID (Entrez) 404037
Content And Storage Store the unopened product at -20 to -80°C
Species Human
Quantity 10 μg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.