missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HAND1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3168-SP
This item is not returnable.
View return policy
Description
HAND1 Polyclonal specifically detects HAND1 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| HAND1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BHLHA27, bHLHa27heart- and neural crest derivatives-expressed protein 1, Class A basic helix-loop-helix protein 27, eHand, Extraembryonic tissues, heart, autonomic nervous system and neural crestderivatives-expressed protein 1, heart and neural crest derivatives expressed 1, Hxt, Thing1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 9421 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| O96004 | |
| HAND1 | |
| E. coli-derived recombinant human HAND1 Met1-Gln215 Accession # O96004 | |
| 25 μg | |
| Primary | |
| Detects human HAND1 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human HAND2 is observed. | |
| Human | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering