missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Hairless Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF5708
This item is not returnable.
View return policy
Description
Hairless Polyclonal specifically detects Hairless in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| Hairless | |
| Polyclonal | |
| Unconjugated | |
| O43593 | |
| HR | |
| E. coli-derived recombinant human Hairless Met1-Thr277 Accession # O43593 | |
| 100 μg | |
| Primary | |
| Detects human Hairless in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| ALUNC, AU, AUHSA277165, FLJ98880, hairless (mouse) homolog, hairless homolog (mouse), HR, MUHH, MUHH1, protein hairless | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 55806 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction