missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HAI-1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 6 publications
Brand: R&D Systems AF1048
This item is not returnable.
View return policy
Description
HAI-1/HGFA Inhibitor 1 Polyclonal specifically detects HAI-1/HGFA Inhibitor 1 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| HAI-1/HGFA Inhibitor 1 | |
| Polyclonal | |
| Unconjugated | |
| NP_003701 | |
| SPINT1 | |
| Mouse myeloma cell line NS0-derived recombinant human HAI-1 Pro37-Glu449 Accession # NP_003701 | |
| 100 μg | |
| Primary | |
| Detects human HAI-1 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse HAI-1 is observed and less than 1% cross-reactivity with recombinant human HAI-2 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| HAI, HAI1, HAI-1, hepatocyte growth factor activator inhibitor 1, Hepatocyte growth factor activator inhibitor type 1, kunitz-type protease inhibitor 1, MANSC2, serine peptidase inhibitor, Kunitz type 1, serine protease inhibitor, Kunitz type 1, SPINT1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6692 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction