missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Guanylyl Cyclase F (NP_001513.2) VersaClone cDNA

Product Code. 18750657
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18750657

Brand: R&D Systems™ RDC3796

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Gene GUCY2F
Accession Number NP_001513.2
Gene ID (Entrez) 2986
Gene Alias CYGF, EC 4.6.1, GC-F, GC-FEC 4.6.1.2, guanylate cyclase 2F, retinalRETGC-2, Guanylate cyclase F, GUC2DL, GUC2Fguanylate cyclase 2D-like, membrane (retina-specific), GUCY2F, RETGC2, retinal guanylyl cyclase 2, Rod outer segment membrane guanylate cyclase 2, ROS-GC2, ROS-GC2RetGC-2
Content And Storage Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.