missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Guanylyl Cyclase C/GUCY2C Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand: R&D Systems MAB2157-100
This item is not returnable.
View return policy
Description
GUCY2C Monoclonal specifically detects GUCY2C in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| GUCY2C | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| DIAR6, EC 4.6.1, GCC, GC-C, guanylate cyclase 2C (heat stable enterotoxin receptor), Guanylate Cyclase C, Guanylyl cyclase C, GUC2C, GUC2CEC 4.6.1.2, GUCY2C, heat-stable enterotoxin receptor, hSTAR, Intestinal guanylate cyclase, MUCIL, STA Receptor, StAR, STARSTA receptor | |
| Human embryonic kidney cell HEK293-derived human Guanylyl Cyclase C/GUCY2C, Ser21-Gln430, Accession # P25092 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 2543C | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 3-25 ug/mL, CyTOF-ready | |
| P25092 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 2984 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction