missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Guanylyl Cyclase C/GUCY2C Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems FAB2157T-100UG
This item is not returnable.
View return policy
Description
GUCY2C Monoclonal specifically detects GUCY2C in Human samples. It is validated for Flow Cytometry.
Specifications
| GUCY2C | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| DIAR6, EC 4.6.1, GCC, GC-C, guanylate cyclase 2C (heat stable enterotoxin receptor), Guanylate Cyclase C, Guanylyl cyclase C, GUC2C, GUC2CEC 4.6.1.2, GUCY2C, heat-stable enterotoxin receptor, hSTAR, Intestinal guanylate cyclase, MUCIL, STA Receptor, StAR, STARSTA receptor | |
| Human embryonic kidney cell HEK293-derived human Guanylyl Cyclase C/GUCY2C, Ser21-Gln430, Accession # P25092 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 2543C | |
| Flow Cytometry 0.25-1 ug/106 cells | |
| P25092 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 2984 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction