missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human GTPBP1 (aa 63-138) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66710 (PA5-66710. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Promotes degradation of target mRNA species. Plays a role in the regulation of circadian mRNA stability. Binds GTP and has GTPase activity.
Specifications
Specifications
| Accession Number | O00178 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 9567 |
| Name | Human GTPBP1 (aa 63-138) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | AL022987; AW045683; C85212; GP1; GP-1; G-protein 1; GTP binding protein 1; GTP-binding protein 1; Gtpbp; Gtpbp1; HSPC018; LOW QUALITY PROTEIN: GTP-binding protein 1 |
| Common Name | GTPBP1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction