missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human GTF3C2 (aa 810-905) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (92%), Rat (92%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57418 (PA5-57418. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Required for RNA polymerase III-mediated transcription. Component of TFIIIC that initiates transcription complex assembly on tRNA and is required for transcription of 5S rRNA and other stable nuclear and cytoplasmic RNAs. May play a direct role in stabilizing interactions of TFIIIC2 with TFIIIC1. [UniProt]
Specifications
Specifications
| Accession Number | Q8WUA4 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 2976 |
| Name | Human GTF3C2 (aa 810-905) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 1300004C11Rik; 2610510G03Rik; AI225816; AU041069; general transcription factor 3 C polypeptide 2; general transcription factor IIIC subunit 2; general transcription factor IIIC, polypeptide 2, beta; general transcription factor IIIC, polypeptide 2, beta 110 kDa; Gtf3c2; KIAA0011; mKIAA0011; TF3C-beta; TFIIIC 110 kDa subunit; TFIIIC110; TFIIIC-BETA; Transcription factor IIIC 110 kDa subunit; transcription factor IIIC subunit beta |
| Common Name | GTF3C2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction