missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GTF2I (aa 431-569) Control Fragment Recombinant Protein

Product Code. 30194085
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30194085

Brand: Invitrogen™ RP102472

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82928 (PA5-82928. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a multifunctional phosphoprotein with roles in transcription and signal transduction. It is deleted in Williams-Beuren syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at chromosome 7q11. 23. Alternative splicing results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 7, 13 and 21.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P78347
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 2969
Name Human GTF2I (aa 431-569) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 6030441I21Rik; BAP135; BAP-135; Bruton tyrosine kinase-associated protein 135; BTKAP1; BTK-associated protein 135; BTK-associated protein, 135 kD; DIWS; Diws1t; general transcription factor II I; general transcription factor IIi; general transcription factor II-I; Gtf2i; Gtf2ird1; GTFII-I; IB291; OTTHUMP00000209509; Spin; SRF-Pho x 1-interacting protein; tfiii; TFII-I; WBS; WBSCR6; Williams-Beuren syndrome chromosomal region 6 protein; Williams-Beuren syndrome chromosome region 6
Common Name GTF2I
Gene Symbol GTF2I
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence FVIKKHELLNSTREDLQLDKPASGVKEEWYARITKLRKMVDQLFCKKFAEALGSTEAKAVPYQKFEAHPNDLYVEGLPENIPFRSPSWYGIPRLEKIIQVGNRIKFVIKRPELLTHSTTEVTQPRTNTPVKEDWNVRIT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt