missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GSTA1 (aa 52-76) Control Fragment Recombinant Protein

Product Code. 30212014
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30212014 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30212014 Supplier Invitrogen™ Supplier No. RP104442

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of a family of enzymes that function to add glutathione to target electrophilic compounds, including carcinogens, therapeutic drugs, environmental toxins, and products of oxidative stress. This action is an important step in detoxification of these compounds. This subfamily of enzymes has a particular role in protecting cells from reactive oxygen species and the products of peroxidation. Polymorphisms in this gene influence the ability of individuals to metabolize different drugs. This gene is located in a cluster of similar genes and pseudogenes on chromosome 6. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08263
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 2938
Name Human GSTA1 (aa 52-76) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 13-hydroperoxyoctadecadienoate peroxidase; Androst-5-ene-3,17-dione isomerase; class alpha; glutathione S-alkyltransferase A1; glutathione S-aryltransferase A1; glutathione S-transferase; glutathione S-transferase 2; glutathione S-transferase A1; Glutathione S-transferase A1, N-terminally processed; glutathione S-transferase A3 subunit; glutathione S-transferase A5; glutathione S-transferase alpha 1; glutathione S-transferase alpha M14; Glutathione S-transferase alpha M14, N-terminally processed; Glutathione S-transferase alpha-3; glutathione S-transferase Ha subunit 1; glutathione S-transferase Ya; Glutathione S-transferase Ya1; glutathione S-transferase Yc-1; glutathione S-transferase Yc1 subunit; glutathione S-transferase, alpha 1 (Ya); Glutathione-S-transferase alpha type (Ya); GST; GST 2-2; GST A3-3; GST AA; GST class-alpha; GST class-alpha member 1; GST HA subunit 1; GST Yc1; GST2; Gst2-1; Gsta; GSTA1; GSTA1-1; Gsta3; Gsta5; GST-epsilon; Gstya; Gstyc1; GTH1; OTTMUSG00000031890; RP11-392A23.1; S-(hydroxyalkyl)glutathione lyase A1; testicular tissue protein Li 80; Yc1
Common Name GSTA1
Gene Symbol GSTA1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence FQQVPMVEIDGMKLVQTRAILNYIA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.