missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GST Omega 1 140A Recombinant Protein

Product Code. 11538891
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11538891 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11538891 Supplier Invitrogen™ Supplier No. RP75564

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

This product contains purified recombinant human GST Omega 1 140A protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number P78417
For Use With (Application) Control, Western Blot
Formulation 20 mM tris HCl with 50% glycerol and no preservative; pH 8
Gene ID (Entrez) 9446
Name Human GST Omega 1 140 A
Quantity 100 μg
Source E. coli
Immunogen N/A
Regulatory Status RUO
Gene Alias epididymis secretory protein Li 21; glutathione S-transferase omega 1; glutathione S-transferase omega 1-1; Glutathione S-transferase omega-1; glutathione-dependent dehydroascorbate reductase; glutathione-S-transferase like; GSTO 1-1; GSTO1; GSTO-1; GSTTLp28; HEL-S-21; MMA(V) reductase; monomethylarsonic acid reductase; P28; S-(Phenacyl)glutathione reductase; SPG-R
Common Name GST Omega 1 140 A
Gene Symbol GSTO1
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Protein Tag His-tag
Sequence Amino acids Met1- Leu241 containing an N-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.