missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GRP94 (aa 282-392) Control Fragment Recombinant Protein

Product Code. 30195500
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30195500

Brand: Invitrogen™ RP102270

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

GRP94 is a 803 amino acid protein belonging to the heat shock protein 90 family. It acts as a molecular chaperone that functions in the processing and transport of secreted proteins. GRP94 and its N-terminal fragment stimulates CTL expansion and maturation of human monocyte-derived dendritic cells (MDDC). It plays a central role in innate as well as acquired immunity, maturation and chemotaxis of dendritic cells, Ab production, cross-priming, as a potential marker in breast cancer and is a peptide acceptor in endoplasmic reticulum and an accessory to peptide loading of MHC class I molecules. Expression of GRP94 suppressed A23187-induced apoptosis and stabilized calcium homeostasis. GRP94 is expressed in melanoma or liver metastases of colon carcinoma cells, human gastric carcinoma BGC-823 cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P14625
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 7184
Name Human GRP94 (aa 282-392) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 108 K heat shock protein; 108 K HSP; 94 kDa glucose-regulated protein; 94 kDa glucose-regulated protein {ECO:0000250; 98 kDa protein kinase; ecgp; endoplasmic reticulum resident protein 99; Endoplasmin; endoplasmin {ECO:0000250; endoplasmin-like protein; endothelial cell (HBMEC) glycoprotein; epididymis luminal protein 35; epididymis secretory sperm binding protein Li 125 m; ERp99; glucose-regulated protein; glucose-regulated protein GRP94; glucose-regulated protein GRP94 precursor; GP96; gp96 homolog; gp96/GRP94; GRP94; GRP-94; heat shock 108 kDa protein; heat shock protein 108; heat shock protein 108.; heat shock protein 90 beta family member 1; heat shock protein 90 kDa beta member 1; heat shock protein 90 kDa beta member 1 {ECO:0000250; heat shock protein 90, beta (Grp94), member 1; heat shock protein 90, beta, member 1; heat shock protein 90 kDa beta (Grp94), member 1; heat shock protein 90 kDa beta family member 1; heat shock protein 90 kDa beta family member 1 L homeolog; heat shock protein 90 kDa beta, member 1; heat shock protein gp96; HEL35; HEL-S-125 m; HSP 108; hsp108; HSP90 beta; hsp90b1; hsp90b1 {ECO:0000250; hsp90b1.L; Polymorphic tumor rejection antigen 1; PPK 98; Ppk 98; a protein kinase; ppk98; stress-inducible tumor rejection antigen gp96; TA-3; Targ2; tra1; Tra-1; transferrin-binding protein; transforming growth factor alpha regulated gene 2; tumor rejection antigen; tumor rejection antigen (gp96) 1; tumor rejection antigen 1; Tumor rejection antigen gp96; UniProtKB:P08113}; XELAEV_18017384mg
Common Name GRP94
Gene Symbol HSP90B1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence WSSKTETVEEPMEEEEAAKEEKEESDDEAAVEEEEEEKKPKTKKVEKTVWDWELMNDIKPIWQRPSKEVEEDEYKAFYKSFSKESDDPMAYIHFTAEGEVTFKSILFVPTS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.