missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GRP78/HSPA5 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB4846
This item is not returnable.
View return policy
Description
GRP78/HSPA5 Monoclonal specifically detects GRP78/HSPA5 in Human samples. It is validated for Western Blot, Simple Western.
Specifications
| GRP78/HSPA5 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
| P11021 | |
| HSPA5 | |
| E. coli-derived recombinant human GRP78 Met1-Leu654 Accession # P11021 | |
| 100 μg | |
| Primary | |
| Detects endogenous human GRP78 at 78 kDa in Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 474421 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 78 kDa glucose-regulated protein, BiP, Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78, FLJ26106, GRP-78, GRP78BIP, Heat shock 70 kDa protein 5, heat shock 70kD protein 5 (glucose-regulated protein, 78kD), heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa), HSP70-5, HSPA5, Immunoglobulin heavy chain-binding protein, MIF2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3309 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction