missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human Growth Hormone R (GHR) Protein, Fc Tag

Product Code. 15816837
Change view
Click to view available options
Quantity:
1 mg
50 μg
Unit Size:
1mg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15816837 1 mg 1mg
15826837 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15816837 Supplier ACROBiosystems Supplier No. GHRH52571MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Growth hormone receptor (GHR) is also known as somatotropin receptor, growth hormone-binding protein (GHBR), which belongs to the type I cytokine receptor family or Type 1 subfamily. GHR contains one fibronectin type-III domain. GHR / GHBR is expressed in various tissues with high expression in liver and skeletal muscle. The soluble form (GHBP) is produced by phorbol ester-promoted proteolytic cleavage at the cell surface (shedding) by ADAM17/TACE. GHR is receptor for pituitary gland growth hormone involved in regulating postnatal body growth. On ligand binding, couples to the JAK2/STAT5 pathway. The soluble form (GHBP) acts as a reservoir of growth hormone in plasma and may be a modulator/inhibitor of GH signaling.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 54.3 kDa. As a result of glycosylation, the protein migrates as 66-85 kDa under reducing (R) condition, and 135-160 kDa under non-reducing (NR) condition (SDS-P

Specifications

Accession Number P10912
Molecular Weight (g/mol) 54.3 kDa
Quantity 1 mg
Source Human GHR, Fc Tag (GHR-H5257) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Tyr 264 (Accession # P10912).
Immunogen Ala 27 - Tyr 264
Storage Requirements -20°C
Gene Alias GHR,GHBP,GH receptor
Common Name Growth Hormone R
Conjugate Unconjugated
Species Human
Protein Tag Fc Tag
Buffer PBS, pH7.4
Endotoxin Level Less than 1.0 EU / μg by the LAL method.
Expression System HEK293
Form Powder
Protein Subtype Bio-Markers and CD Antigens
Purity or Quality Grade >95% as determined by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.