missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Growth Hormone Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1067-100
This item is not returnable.
View return policy
Description
Growth Hormone Monoclonal specifically detects Growth Hormone in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| Growth Hormone | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Neutralization 2-8 ug/mL | |
| CAA23779 | |
| GH1 | |
| E. coli-derived recombinant human Growth Hormone Phe27-Phe217 Accession # CAA23779 | |
| 100 μg | |
| Primary | |
| Detects human Growth Hormone (GH) in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant rat GH is observed. | |
| Human | |
| Supernatant |
| Western Blot, Neutralization | |
| 178902 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| GH1, GHB5, GHIGHD1B, GHN, GH-N, GHNGrowth hormone, GH-Nsomatotropin, growth hormone 1Pituitary growth hormone, hGH-N, IGHD1A, IGHD2, Somatotropin | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2688 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction