missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GRK5 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4539
This item is not returnable.
View return policy
Description
GRK5 Monoclonal specifically detects GRK5 in Human samples. It is validated for Immunocytochemistry.
Specifications
| GRK5 | |
| Monoclonal | |
| LYOPH | |
| Immunocytochemistry 8-25 ug/mL | |
| EC 2.7.11, EC 2.7.11.16, FLJ39780, G protein-coupled receptor kinase 5, G protein-coupled receptor kinase GRK5, GPRK5, GPRK5FP2025 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human GRK5 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) GRK6 is observed and no cross-reactivity with rhGRK4 is observed. | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 691816 | |
| Unconjugated | |
| P34947 | |
| GRK5 | |
| E. coli-derived recombinant human GRK5 Pro463-Ser590 Accession # P34947 | |
| 100 μg | |
| 2869 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction