missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GRIK5 (aa 210-297) Control Fragment Recombinant Protein

Product Code. 30193982
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30193982

Brand: Invitrogen™ RP108628

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. Grik5, also known as kainate-preferring glutamate recptor subunit KA2, belongs to the kainate family of glutamate receptors, which are composed of four subunits and function as ligand-activated ion channels. Grik5 is highly homologous to the related ionotrophic glutamate receptor Grik4 (also known as KA1). Like Grik4, Grik5 does not form homomeric channels, but instead forms heteromers with Grik2. In Grik2- but not Grik1-null mice, Grik5 surface expression is greatly reduced in neurons, indicating that Grik2/Grik5 heteromers are required for exit from the endoplasmic reticulum to the cell surface.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q16478
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 2901
Name Human GRIK5 (aa 210-297) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias EAA2; Excitatory amino acid receptor 2; GluK5; GluR gamma-2; GluRgamma2; glutamate ionotropic receptor kainate type subunit 5; glutamate receptor; glutamate receptor gamma-2; Glutamate receptor ionotropic, kainate 5; glutamate receptor KA2; glutamate receptor KA-2; glutamate receptor, ionotropic kainate 5; glutamate receptor, ionotropic, kainate 5; glutamate receptor, ionotropic, kainate 5 (gamma 2); GRIK2; GRIK5; iGlu5; KA2; LOW QUALITY PROTEIN: glutamate receptor ionotropic, kainate 5; MGC128958 protein
Common Name GRIK5
Gene Symbol GRIK5
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KVSTIIIDANASISHLILRKASELGMTSAFYKYILTTMDFPILHLDGIVEDSSNILGFSMFNTSHPFYPEFVRSLNMSWRENCEASTY
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.