missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Grik3 Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Grik3 Synthetic Peptide, PEP-0567, from Invitrogen.
Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. This gene product belongs to the kainate family of glutamate receptors, which are composed of four subunits and function as ligand-activated ion channels. It is not certain if the subunit encoded by this gene is subject to RNA editing as the other 2 family members (GRIK1 and GRIK2). A Ser310Ala polymorphism has been associated with schizophrenia, and there are conflicting reports of its association with the pathogenesis of delirium tremens in alcoholics.
Specifications
Specifications
| Gene ID (Entrez) | 2899 |
| Species | Human |
| Purification Method | purified |
| Accession Number | Q13003 |
| Common Name | Human Grik3 |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction