missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human GRHL3 (aa 50-135) Control Fragment Recombinant Protein

Product Code. 30203050
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30203050

Brand: Invitrogen™ RP106979

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (84%), Rat (84%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66299 (PA5-66299. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the grainyhead family of transcription factors. The encoded protein interacts with leader-binding protein 32 (LBP-32) and brother of mammalian grainyhead (BOM), and may function as a transcription factor during development. Multiple transcript variants encoding distinct isoforms have been identified for this gene. Additional transcript variants have been described, but their biological nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8TE85
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 57822
Name Human GRHL3 (aa 50-135) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AI561912; ct; Get1; grainy head-like 3; grainyhead like transcription factor 3; grainyhead-like 3; grainyhead-like 3 (Drosophila); grainyhead-like protein 3 homolog; Grhl3; Sister of mammalian grainyhead; sister-of-mammalian grainyhead; sister-of-mammalian grainyhead protein; SOM; TFCP2L4; Transcription factor CP2-like 4; transcription factor hSOM1; VWS2
Common Name GRHL3
Gene Symbol GRHL3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VNGDDDSVAALSFLYDYYMGPKEKRILSSSTGGRNDQGKRYYHGMEYETDLTPLESPTHLMKFLTENVSGTPEYPDLLKKNNLMSL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.