missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human Gremlin / GREM1 Protein, His Tag

Product Code. 15846897
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15846897 100 μg 100µg
15856897 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15846897 Supplier ACROBiosystems Supplier No. GR1H52H3100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gremlin is also known as Cysteine knot superfamily 1, BMP antagonist 1 (CKTSF1B1), DAN domain family member 2 (DAND2), Down-regulated in Mos-transformed cells protein (DRM), Increased in high glucose protein 2 (IHG-2), Cell proliferation-inducing gene 2 protein (PIG2) or Gremlin-1 (GREM1), which is highly expressed in small intestine, fetal brain and colon. Gremlin / GREM-1 interacts with SLIT1 and SLIT2 in a glycosylation-dependent manner. Gremlin may play an important role during carcinogenesis and metanephric kidney organogenesis, as a BMP antagonist required for early limb outgrowth and patterning in maintaining the FGF4-SHH feedback loop. Gremlin down-regulates the BMP4 signaling in a dose-dependent manner and acts as inhibitor of monocyte chemotaxis.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 20.2 kDa. The protein migrates as 28-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.

Specifications

Accession Number NP_037504
Molecular Weight (g/mol) 20.2 kDa
Quantity 100 μg
Source Human Gremlin, His Tag (GR1-H52H3) is expressed from human 293 cells (HEK293). It contains AA Lys 25 - Asp 184 (Accession # NP_037504).
Immunogen Lys 25 - Asp 184
Storage Requirements -20°C
Gene Alias GREM1,CKTSF1B1,DAND2,DRM,PIG2
Common Name Gremlin
Conjugate Unconjugated
Species Human
Protein Tag His Tag
Buffer PBS, pH7.4 with 1 mM EDTA
Endotoxin Level Less than 1.0 EU / μg by the LAL method.
Expression System HEK293
Form Powder
Protein Subtype Cytokines and Growth Factors
Purity or Quality Grade >95% as determined by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.