missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human Gremlin-1 Recombinant Protein, PeproTech®
Click to view available options
Quantity:
10 μg
100 μg
2 x 500 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
500µg
50µg
Pack of 2
Description
120-42-1MG will be provided as 2 x 500 μg (120-42-500UG). Recombinant Human Gremlin-1 is a 18.3 kDa protein containing 160 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
In Xenopus, Gremlin belongs to a novel gene family that act as BMP antagonists in embryonic explants. The human homolog of Drm/Gremlin (CKTFS1B1) is localized on human chromosome 15q13--> q15. DRM-specific mRNA is expressed in different human tissues, including brain, ovary, intestine and colon. It has been suggested that down-regulation of DRM is associated with tumor progression, and support the hypothesis that human DRM may play an important role during both neuroembryological development and carcinogenesis.
Specifications
Specifications
| Accession Number | O60565 |
| For Use With (Application) | Control, Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 26585 |
| Molecular Weight (g/mol) | 18.3 kDa |
| Name | Human Gremlin-1 |
| Purification Method | Purified |
| Quantity | 10 μg |
| Source | CHO cells |
| Storage Requirements | -20°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction