missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GRAP2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4640
This item is not returnable.
View return policy
Description
GRAP2 Monoclonal specifically detects GRAP2 in Human samples. It is validated for Western Blot.
Specifications
| GRAP2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| O75791 | |
| GRAP2 | |
| E. coli-derived recombinant human GRAP2 Met1-Arg330 Accession # O75791 | |
| 100 μg | |
| Primary | |
| Detects endogenous human GRAP2 in Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 475804 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Adapter protein GRID, GADS, GADSSH3-SH2-SH3 adapter Mona, GRB2L, GRB-2-like protein, GRB2-related adapter protein 2, GRB2-related adaptor protein 2, GRB2-related protein with insert domain, GRBLG, GRBLGGRAP-2, GrbX, Grf40, Grf-40, Grf40 adapter protein, GRID, GRIDGRPL, Growth factor receptor-binding protein, growth factor receptor-bound protein 2-related adaptor protein 2, GRPL, Hematopoietic cell-associated adapter protein GrpL, hematopoietic cell-associated adaptor protein GRPL, Mona, P38, Protein GADS, SH3-SH2-SH3 adaptor molecule | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9402 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction