Learn More
Human GPVI Antibody, R&D Systems™
Sheep Polyclonal Antibody has been used in 1 publication
Brand:  R&D Systems AF3627
Additional Details : Weight : 0.00970kg
 View more versions of this product
Product Code. 18012422
  • £469.23 / 100µg
Estimated Shipment: 04-06-2024
to see stock.
Description
Description
GPVI Polyclonal specifically detects GPVI in Human samples. It is validated for Western Blot.
Specifications
Specifications
GPVI
Polyclonal
Unconjugated
Q9HCN6
GP6
Mouse myeloma cell line NS0-derived recombinant human GPVI (R&D Systems, Catalog # 3627-GP) Gln21-Lys267 Accession # Q9HCN6
100 μg
Primary
Detects human GPVI in direct ELISAs and Western blots.
Human
IgG
Western Blot
LYOPH
Western Blot 0.1 ug/mL
Glycoprotein 6, glycoprotein VI (platelet), GP6, GPIV, GPVIplatelet collagen receptor, MGC138168, platelet glycoprotein VI
Sheep
Affinity Purified
RUO
51206
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.