missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GPRC5D Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB63001-100
This item is not returnable.
View return policy
Description
GPRC5D Monoclonal specifically detects GPRC5D in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| GPRC5D | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| G protein-coupled receptor, family C, group 5, member D, G-protein coupled receptor family C group 5 member D, MGC129713, MGC129714, orphan G-protein coupled receptor | |
| NS0 mouse myeloma cell line transfected with human GPRC5D, Met1-Val359, Accession # Q9NZD1 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 571932 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Q9NZD1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 55507 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction