missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human GPR89A (NP_001091081.1) VersaClone cDNA
Specifications
Specifications
| Gene | GPCR GPR89 |
| Accession Number | NP_001091081.1 |
| Gene ID (Entrez) | 653519 |
| Gene Alias | FLJ10492, FLJ11696, FLJ36909, FLJ79270, G protein-coupled receptor 89 A, Golgi pH regulator A, GPHR, GPHRA, GPR89, GPR89B, MGC688, MGC87324, protein GPR89, Protein GPR89A, Putative MAPK-activating protein PM01, Putative NF-kappa-B-activating protein 90, SH120, UNQ192 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?